Learn More
Fisherbrand™ Traceable™ Silicone Pump Tubing
Ideal for use with food, pharmaceuticals, and other critical solutions
 View more versions of this product
Catalog No. 15-078-270
Add to cart
Description
Description
Ideal for use with food, pharmaceuticals, and other critical solutions
  • Replacement tubing for Variable-Speed Pump
  • Can be sterilized by autoclaving
Specifications
Specifications
Autoclavable
6.35 mm
25 ft.
Pump Tubing
1/4 in.
Silicone
7.6 m
Medium flow and Medium/High Flow Variable-Speed Pump