Control Company Traceable™ Silicone Pump Tubing
Ideal for use with food, pharmaceuticals, and other critical solutions
 View more versions of this product
Catalog No. 15-078-271
  Check Availability
Add to Cart
    Specifications
    Diameter (English) Inner 1/16 in.
    For Use With (Equipment) All Variable-Speed pumps
    Length (Metric) 7.6m
    Autoclavable Yes
    Length (English) 25 ft.
    Material Silicone
    • Replacement tubing for Variable-Speed Pump
    • Can be sterilized by autoclaving
  • Description & Specifications
Related Products
Product Content Feedback