Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Fisher Scientific™ Traceable™ Temperature Chart Recorders Available on GSA/VA Contract for Federal Government customers only.

Provides a permanent record, which is easy to read, duplicate, fax, scan and file. Fisher Scientific Traceable Temperature Chart Recorders are single-use, disposable recorders for monitoring temperatures in refrigerators, supply rooms, food storage, chemical warehouses, animal quarters, incubators, goods-in-transit, walk-in chambers, biological materials, and manufacturing plants.

 View more versions of this product

Catalog No. 14-648-40


  Check Availability
Add to cart

Specifications

Description

  • Shock-resistant ABS plastic case
  • Eliminate messy pens, expensive instrumentation, confusing software and training
  • Display temperature in both °F and °C on large 5.5 in. dia. (14cm) chart
  • Battery-operated motor turns the chart with accuracy of ±1 second per day
  • Easy-to-read chart shows temperature and time/day of occurrence
  • Transparent window on recorder allows viewing of current temperature and history
  • Records for 5, 10 or 40 days
  • Chart, marker, and battery are preloaded—simply pull tab to begin recording
  • Tamper-evident seal maintains integrity of unit and results
  • Case measures 3.87L x 2.5W x 5.25 in.H (13.3 x 9.8 x 6.4cm)

Order Info

  • Weight: 11 oz. (312g)
  • Case measures 5.25L x 3.87W x 2.5 in.H (13.3 x 9.8 x 6.4cm)

Certifications

Individually serial-numbered Traceable™ Certificate is provided from an ISO 17025 calibration laboratory accredited by A2LA. It indicates traceability to standards provided by NIST.

Description & Specifications

Specifications

±1°C/±2°F
AA Alkaline
-20° to 100°F
6.4cm
5 Days
-28° to 38°C
312g
9.8cm
ISO 17025, NIST
5-Day Recorder
2.5 in.
One "AA" Alkaline battery
Bimetallic
11 oz.
3.87 in.
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit