Learn More
Fisherbrand™ Traceable™ Tubing Sets
For use with Variable-Speed Pumps
Description
For use with Variable-Speed Pumps
- Replacement set of tubing and fittings/nipples
- Identical to the set supplied with pump
- Supplied with 1/50, 1/16, 3/32, 3/16 and 1/4 in. I.D. tubing and corresponding fittings/nipples
Specifications
Tubing Set | |
1/50, 1/16, 3/32, 3/16, 1/4 in. | |
Ultra Low Flow and Low Flow Variable-Speed Pump |
Tubing Replacemant Set | |
0.508, 1.58, 2.38, 4.76, 6.35 mm |