Learn More
Fisherbrand™ Traceable™ Tubing Sets, For Ultra Low Flow and Low Flow Variable-Speed Pump
For use with Variable-Speed Pumps
$78.30 - $80.50
Specifications
Description Tubing Replacemant Set
View More Specs
Products 3
Catalog Number For Use With Price Quantity & Availability  
Catalog Number For Use With Price Quantity & Availability  
15-078-274
View Documents
Ultra Low Flow and Low Flow Variable-Speed Pump
Each for $78.30
Add to cart
15-078-278
View Documents
Medium Flow Variable-Speed Pump
Each for $80.50
Add to cart
15-078-279
View Documents
Medium/High Flow Variable-Speed Pump
Each for $80.50
Add to cart
Description
Description
For use with Variable-Speed Pumps
  • Replacement set of tubing and fittings/nipples
  • Identical to the set supplied with pump
  • Supplied with 1/50, 1/16, 3/32, 3/16 and 1/4 in. I.D. tubing and corresponding fittings/nipples
Specifications
Specifications
Tubing Replacemant Set