Learn More
Fisherbrand™ Traceable™ Workstation Barometer with Clock
Ideal for recording ambient changes in the lab, monitoring conditions affecting sample analysis and assisting in the prediction of weather changes
Catalog No. 15-079-610
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
Description
Description
Fisher Scientific™ Traceable™ Workstation Barometer with Clock is ideal for recording ambient changes in the lab, monitoring conditions affecting sample analysis and assisting in the prediction of weather changes.
  • Measures barometric pressure and graphically displays the barometric trend
  • Displays temperature and time of day/date
  • Adjustable altitude compensation from -900 to 3600 feet (-300 to 1200 meters)
  • Graphic barometric trend shows present reading and previous one-, three-, six- and 12-hour readings in a bar chart format
  • Forecasting indicators show weather conditions for the next 12-hours using symbols for sunny, cloudy and rainy
  • Clock shows time in AM/PM or 24-hour format and date
  • Barometric range: 23.62 to 31.01 inHg / 800 to 1050 mbar; resolution: 0.063 inHg / 1 mbar; accuracy: ±6 mb (890 to 1020 mb); otherwise ±10 mb
  • Temperature range: -9 to 70°C (-22 to 158°F); Resolution: 0.5°; Accuracy: ±1°C (between 32 to 104°F/0 to 40°C); otherwise ±2°C
  • May be wall mounted
Certifications
Individually serial-numbered Traceable™ Certificate is provided from our ISO 17025 calibration laboratory accredited by A2LA™.
Specifications
Specifications
±6 mbar (890 to 1020 mb); otherwise ±10 mb
−9.5 to 70.5°C
7.62 cm
7.75 in.
2.5 in.
0.27 kg
0.5°C
Workstation Barometer with Clock
3 in.
19.69 cm
6.35 cm
8 oz.