Learn More
Fisher Scientific Universal Spill Kit
Small enough to fit on a lab bench.
$98.00
Specifications
For Use With (Application) | Safe for use on all chemicals except hydrofluoric acid |
---|
Alert:
These products contain natural rubber latex, a component of which may cause allergic reaction in some users. Use only as directed and in accordance with manufacturer instructions and warnings. Please consult your institution's policies regarding use of these products.
Sorbent and pillows not suitable for use with hydrofluoric acid.
Includes: One carton of super sorbent; six pillows; two pads; one pair each of goggles, nitrile gloves and latex boots; one disposal bag; universal instruction sheet.
Catalog Number | Mfr. No. | Capacity | Product Type | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Capacity | Product Type | Price | Quantity & Availability | ||||
S47441
![]() ![]() |
Fisher Scientific
S47441 |
2.5 gal., 10L | Universal Spill Kit |
Each for $98.00
|
|
Description
- Universal sorbent eliminates guesswork safe for use on all chemicals except hydrofluoric acid
- Kit absorbs 2.5 gal. (10L)
- Contains 2.5 lb. (1.1kg) carton of universal sorbent
- Each 4 x 14 x 1 in. (10.2 x 35.6 x 2.5cm) lab pillow (also available separately) absorbs 0.22 gal. (750mL)
Specifications
Safe for use on all chemicals except hydrofluoric acid |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.