Fisher Scientific™ Wellwash™ Microplate Washers Available on GSA/VA Contract for Federal Government customers only.

Easy-to-use microplate washers for ELISA applications

 View more versions of this product

Catalog No. 14430201

 N/A
N/A

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000. 


    Specifications

    Plate Type 96-Well Plates
    Cell Washing No
    Display LCD
    Width (English) 13.6 in.
    Height (English) 9.4 in.
    Weight (Metric) 9kg
    Wash/Waste Bottle Capacity 1
    Weight (English) 20 lb.
    Includes 1 x 2L wash bottle, 1 x 2L waste bottle, 1 x 8 wash head, aerosol cover (Wellwash); 2 x 2L wash bottles, 1 x 2L rinse bottle, 1 x 4L waste bottle, 2 x 8 wash head, aerosol cover (Wellwash Versa)
    Wash Heads 1 x 8; 1 x 12
    Volume (Metric) Dispense 50 to 400μL
    Volume (Metric) Prime 50 to 1000μL
    Volume (Metric) Residual <1.5μL
    Certifications/Compliance CE Marked
    For Use With (Application) ELISA-Based Assays; Cell Washing
    Volume (Metric) Rinse 50 to 1000μL
    Volume (Metric) Wash 50 to 1000μL
    Width (Metric) 34.5cm
    Depth (Metric) 38.5cm
    Depth (English) 15.2 in.
    Height (Metric) 24cm
    Type Wellwash Versa

  • Description & Specifications

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit