Learn More
Fisherbrand Wellwash and Wellwash Versa Microplate Strip Washer: Bottles
Accessories for Fisher Scientific Wellwash and Wellwash Versa Microplate Strip Washer
Description
For use with Fisher Scientific Wellwash and Wellwash Versa Microplate Strip Washer
- Subsitute bottles of different sizes for added convenience
- Wash and waste bottles include liquid level sensors
- 4L wash and 9L waste bottles available for automation applications
ELISA-based assays, cell washing
Specifications
135.25 oz. | |
135.25 oz., 4L | |
Wash Bottle A | |
Cap with tubing and liquid level sensors |
4L | |
Wellwash™ and wellwash™ versa™ microplate strip washer | |
Round |