Learn More
Fisherbrand™ Wellwash™ and Wellwash™ Versa™ Microplate Strip Washer: Bottles
Accessories for Fisher Scientific Wellwash and Wellwash Versa Microplate Strip Washer
 View more versions of this product
Catalog No. 14430216
Add to cart
Description
Description
For use with Fisher Scientific Wellwash and Wellwash Versa Microplate Strip Washer
  • Subsitute bottles of different sizes for added convenience
  • Wash and waste bottles include liquid level sensors
  • 4L wash and 9L waste bottles available for automation applications
ELISA-based assays, cell washing
Specifications
Specifications
304.32 oz.
304.32 oz., 9L
Waste Bottle
Cap with tubing and liquid level sensors
9L
Wellwash™ versa™ microplate strip washer
Rectangular