Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Fisherbrand™ Wellwash™ and Wellwash™ Versa™ Microplate Strip Washer: Spare Bottles

Accessories for Fisher Scientific Wellwash and Wellwash Versa Microplate Strip Washer

 View more versions of this product

Catalog No. 14430223


Add to cart

Description

Description

For use with Fisher Scientific Wellwash and Wellwash Versa Microplate Strip Washer

Subsitute bottles of different sizes for added convenience

ELISA-based assays, cell washing

Specifications

Specifications

67.62 oz.
67.62 oz., 2L
Storage Bottle
2L
Wellwash™ and wellwash™ versa™ microplate strip washer
Solid cap
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit