Learn More
Fisherbrand™ Workstation-Traceable™ Radio-Controlled Clock
SCI_ED
Records date, day, and temperature from a direct signal from the National Institute of Standards and Technology.
Catalog No. S95152PROMO
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
Description
Description
Ideal for recording the receiving of samples, noting the exact time an event occurred, and supporting documentation.
  • Signal automatically adjusts for A.M./P.M., 24-hour time, day/date, daylight saving time, leap year, and leap seconds
  • Backlighting permits viewing in total darkness
  • Alarm volume increases until turned off
  • LCD 0.5 in. (13mm) digits readable from more than 10 ft. (4.8m)
  • One “AA” battery with one-year continuous-use life span included
Certifications
Individually serial-numbered Traceable™ Certificate is provided from an ISO 17025 calibration laboratory accredited by A2LA™. It indicates traceability to standards provided by NIST™.