Learn More
Fisherbrand™ Benchtop Beta-Radiation Shields
Maximum β-radiation protection
 View more versions of this product
Catalog No. FB-BT1FIS
Add to cart
Alert:
Organic solvents such as ethanol may cause acrylic to craze or crack.
Description
Description
Protect personnel from β-emitters used in biological research including 32P, 35S, 14C and 3H, while allowing a full view of work.
  • Transparent, 0.5 in. thick (1.3cm), high-quality acrylic exceeds the 0.38 in. thick (1.0cm) standard for maximum protection
  • Ergonomic design keeps samples easily accessible—minimizes spills, protects against bloodborne pathogens and other infectious agents
  • Upright design is ideal for use with chromatography columns
  • Angled shield with large base holds block or microtube racks
  • Clean with warm water and a mild detergent to remove debris and maintain visibility
Specifications
Specifications
Beta Radiation Shield
31cm
Maximum beta-radiation protection
15cm
18.5 in.
0.5 in.
12 in.
6 x 12 x 18.5 in. (15 x 31 x 47cm)
6 in.
Acrylic
47cm
1.27cm