Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Fisherbrand™ Gel Combs for Horizontal Gel Electrophoresis Systems Products for Science Education

A wide variety of accessory combs for horizontal gel electrophoresis systems.

$87.00

Specifications

For Use With (Equipment) Owl™ A1 Large Gel Electrophoresis System
Type Standard Comb
Length (English) Gel 9.8 in.
Length (Metric) Gel 25 cm
Width (English) Gel 7.8 in.
View More Specs

Products 1
Catalog Number Teeth Count Width (English) Teeth Price Quantity & Availability  
Catalog Number Teeth Count Width (English) Teeth Price Quantity & Availability  
S98626 Products for Science Education View Documents 20 0.17 in. Each for $87.00
Add to cart
 
Specifications

Specifications

Owl™ A1 Large Gel Electrophoresis System
9.8 in.
7.8 in.
Standard Comb
25 cm
20 cm
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit