Learn More
Fisherbrand™ Intensifying Screens
Decrease exposure times and lower levels of radioactivity needed for autoradiography of gels
 View more versions of this product
Catalog No. TFIS1417
Add to cart
This item is not returnable. View return policy
Description
Description
Particles emitted from radioactively labeled samples in gel pass through film and strike intensifying screen. Inorganic phosphors absorb energy, converting it to ultraviolet or blue light, which then exposes the film.
  • For use with isotopes that emit high-energy beta particles (e.g., 32P, 45Ca) or gamma particles (e.g., 125I)
  • Fit autoradiography cassettes for common film sizes
Specifications
Specifications
Intensifying Screen
14 in.
17 in.
14 x 17 in. (355.6 x 431.8 mm)
355.6 mm
431.8 mm