Learn More
Fisherbrand™ Intensifying Screens
Decrease exposure times and lower levels of radioactivity needed for autoradiography of gels
 View more versions of this product
Catalog No. TFIS57
This item has been discontinued and is no longer available. Please call customer service for assistance: 1-800-766-7000.
Description
Description
Particles emitted from radioactively labeled samples in gel pass through film and strike intensifying screen. Inorganic phosphors absorb energy, converting it to ultraviolet or blue light, which then exposes the film.
  • For use with isotopes that emit high-energy beta particles (e.g., 32P, 45Ca) or gamma particles (e.g., 125I)
  • Fit autoradiography cassettes for common film sizes