FisherBiotech™ MicroProbe™ Manual Staining System
Performs multistep staining protocols and simplifies standard chemical staining, immunostaining, and in situ hybridization
Compact system stains 20 slides at a time using capillary gap technology.
Includes: MicroProbe Staining Station with eight reagent buckets and lids; Locator tray for Isolon holders; Four Isolon holders; Five 30-well rubber reagent isolators; Five 3-well rubber reagent isolators; Six absorbent pads; slide holder with 72 ProbeOn microscope slides; MicroProbe Incubator with lid, calibration block, and power cord; User manual; Application guide
  • Products 1
  • Description & Specifications
Products
Catalog Number Description Price Quantity Availability  
15-188-10
Configured for 120/240V, 50/60Hz
N/A N/A N/A  
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000. 
Descriptions & Specifications
Related Products