Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Fisherbrand™ Vacuum Systems for Gel Drying Systems

For quick gel drying

 View more versions of this product

Catalog No. FB-GDP-X10

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.


Includes: Pump only

Description

Description

Dual pumping action works with the heat from the gel dryer to dry gels fast. Hot vapors from the gel dryer are condensed and isolated from the rest of the system by a room-temperature vapor pump. A compact, high-efficiency dry vacuum pump provides the deep vacuum needed to get the last liquid out of the gel. The two-pump system works as quickly as a two-stage vacuum pump and cold trap, and faster than a conventional stand-alone diaphragm pump or water jet pump. The trapping medium doesn't mix with the condensate, so it can be reused.

  • As supplied with FB-GD-X1 Systems
  • Work with most gel dryers, including Bio-Rad™, Hoefer Scientific, and Labconco™

Certifications

CE

Specifications

Specifications

115V
CE
15.5 x 11.15 x 13.6 in. (39.4 x 28.3 x 34.5cm)
11.5L/min.
60Hz
34.5cm
39.4cm
28.3cm
Pump only
115V 60Hz
U.S. Standard
Gel drying system
13.6 in.
15.5 in.
11.15 in.
Documents
Accessories
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit