Learn More
Fisherbrand™ Vertical Electrophoresis Casters
Cast up to four gels at once, each in its own pouch
 View more versions of this product
Catalog No. FB-GC16-1
Add to cart
Includes: Stand, 25 Gel pouches, Spacer placer and blocking plate, Three-point leveling system and built-in bubble level, Nonskid rubber feet
Description
Description
  • Casters use a patented procedure
  • Fitted pouch holds acrylamide during polymerization
  • Each pouch acts as the bottom spacer for quick, easy casting
  • Pouches are heat-sealable for gel storage
  • Casters are available in three sizes to match FisherBiotech Vertical Electrophoresis Systems or similar systems
Order Info
Additional accessories available; contact your Customer Service Representative for more information.
Specifications
Specifications
6 in.
16cm
P9 Vertical Electrophoresis System
Stand, 25 gel pouches, spacer placer and blocking plate, three point leveling system and built-in bubble level, Nonskid Rubber feet
10cm
15.24cm
19.05cm
Vertical Caster
3.9 in.