Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Fisherbrand™ 0.2mL PCR Tube Strips Available on GSA/VA Contract for Federal Government customers only.

Ideal for use in 0.2mL, 96-well V-bottom thermal cyclers

 View more versions of this product

Catalog No. 14-230-219


Add to cart

Description

Description

Ideal for use in 0.2mL, 96-well V-bottom thermal cyclers

  • 0.25mL maximum tube volume when capped
  • Strips of eight tubes
  • Can be cut into individual tubes
  • Thin-wall design
  • Cleanroom produced
  • Virgin polypropylene
  • Available with flat cap strips, domed cap strips, or without cap strips

This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.

Certifications

Certified RNase-, DNase-, DNA- and pyrogen-free

Specifications

Specifications

Thin Wall 8-Strip PCR Tube
Human DNA, RNase, DNase, endotoxins free
Polypropylene
250 Case
0.2mL 96-well V-bottom thermal cyclers, PCR and qPCR/Real-Time PCR applications
Flat
Yellow
0.2mL
Strip
0.2mL
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit