Learn More
Fisherbrand Colored Labeling Tape
Colored Label Tape is useful for many labeling applications. It can be written on and will adhere to most surfaces.
$49.40 - $103.50
Specifications
Format | Tape |
---|---|
Product Type | Colored Label Tape |
Quantity | 4/Pk. |
For Use With (Application) | Waterproof, Oil and Acid resistant |
Material | Paper |
Catalog Number | Color | Length (English) | Length (Metric) | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Color | Length (English) | Length (Metric) | Price | Quantity & Availability | ||||
15-901-5B
![]() |
Yellow | 14 yd. | 13 m |
Pack of 4 for $49.40
Case of 4 PK for $166.90 |
![]() |
15-901-5A
![]() |
White | 14 yd. | 13 m |
Pack of 4 for $49.40
Case of 4 PK for $166.90 |
![]() |
15-901-5C
![]() |
Green | 14 yd. | 13 m |
Pack of 4 for $49.40
Case of 4 PK for $166.90 |
![]() |
15-901-5D
![]() |
Pink | 14 yd. | 13 m |
Pack of 4 for $49.40
Case of 4 PK for $166.90 |
![]() |
15-901-5J
|
Copper | 14 yd. | 13 m |
Pack of 4 for $49.40
Case of 4 PK for $166.90 |
|
15-901-5G
![]() |
Blue | 14 yd. | 13 m |
Pack of 4 for $49.40
Case of 4 PK for $166.90 |
![]() |
15-901-5H
![]() |
Aqua | 14 yd. | 13 m |
Pack of 4 for $49.40
Case of 4 PK for $166.90 |
![]() |
15-901-5F
![]() |
Orange | 14 yd. | 13 m |
Pack of 4 for $49.40
Case of 4 PK for $166.90 |
![]() |
15-901-5K
![]() |
Gray | 14 yd. | 13 m |
Pack of 4 for $49.40
Case of 4 PK for $166.90 |
![]() |
15-901-5L
![]() |
Lavender | 14 yd. | 13 m |
Pack of 4 for $49.40
Case of 4 PK for $166.90 |
![]() |
15-901-5E
![]() |
Red | 14 yd. | 13 m |
Pack of 4 for $49.40
Case of 4 PK for $166.90 |
![]() |
15-901-5M
|
Lime Green | 14 yd. | 13 m |
Pack of 4 for $49.40
Case of 4 PK for $166.90 |
|
15-901-5Q
|
Violet | 14 yd. | 13 m |
Pack of 4 for $49.40
Case of 4 PK for $166.90 |
|
15-901-5N
|
Rose | 14 yd. | 13 m |
Pack of 4 for $49.40
Case of 4 PK for $166.90 |
|
15-901-20H
![]() |
Aqua | 60 yd. | 55 m |
Case of 4 for $103.50
|
|
15-901-20J
![]() |
Copper | 60 yd. | 55 m |
Case of 4 for $103.50
|
|
15-901-20K
![]() |
Gray | 60 yd. | 55 m |
Case of 4 for $103.50
|
|
15-901-20M
![]() |
Lime Green | 60 yd. | 55 m |
Case of 4 for $103.50
|
|
15-901-20L
![]() |
Lavender | 60 yd. | 55 m |
Case of 4 for $103.50
|
|
15-901-20N
![]() |
Rose | 60 yd. | 55 m |
Case of 4 for $103.50
|
|
15-901-20Q
![]() |
Violet | 60 yd. | 55 m |
Case of 4 for $103.50
|
|
15-901-20G
![]() |
Blue | 60 yd. | 55 m |
Case of 4 for $103.50
|
|
15-901-20A
![]() ![]() |
White | 60 yd. | 55 m |
Case of 4 for $103.50
|
![]() |
15-901-20B
![]() ![]() |
Yellow | 60 yd. | 55 m |
Case of 4 for $103.50
|
![]() |
15-901-20E
![]() ![]() |
Red | 60 yd. | 55 m |
Case of 4 for $103.50
|
![]() |
15-901-20F
![]() ![]() |
Orange | 60 yd. | 55 m |
Case of 4 for $103.50
|
![]() |
15-901-20C
![]() ![]() |
Green | 60 yd. | 55 m |
Case of 4 for $103.50
|
![]() |
15-901-20D
![]() |
Pink | 60 yd. | 55 m |
Case of 4 for $103.50
|
|
Description
Colored Label Tape is useful for many labeling applications. It can be written on and will adhere to most surfaces.
- Strong, durable, and oil-, water-, and acid-resistant
- Excellent for color coding
- Withstand temperatures from -23° to +121°C (-10° to +250°F)
- Can be written on with pen, pencil, or permanent marker
- Stick to most surfaces including glass, plastic and metal
- Remove cleanly and easily
Tapes are supplied without dispenser.
This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.
Specifications
Tape | |
4/Pk. | |
Paper | |
-23°C to +121°C |
Colored Label Tape | |
Waterproof, Oil and Acid resistant | |
-10 to 250°F | |
0.75 in. |
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.