Fisherbrand™ 100-Place Polypropylene Storage Boxes

100-place polypropylene storage boxes provide compact, safe storage for 0.5, 1.5 and 2.0mL tubes and cryo vials. Unlike cardboard boxes they will not break down if exposed to moisture or heavy use

 View more versions of this product

Catalog No. 14-100-B

  Check Availability
Add to Cart

    Specifications

    Color Blue
    For Use With (Application) Provide compact and safe storage for 0.5, 1.5 and 2.0ml tubes and cryo vials
    For Use With (Equipment) 0.5, 1.5 and 2.0mL tubes and cryo vials
    Includes Clear translucent hinged lids with snap tab, alphanumeric grid label
    No. of Places 100.0
    Temperature Range (English) -112° to 249.8°F
    Temperature Range (Metric) -80° to 121°C
    Material Polypropylene
    Product Type Cryo Storage Box
    Quantity 5 Pack

  • Description & Specifications

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit