Learn More
Fisherbrand™ Octagon Stir Bars
Octagon magnetic stirring bars with integral pivot ring are the most commonly used shape. Choose from red, yellow or blue.
Catalog No. 14-512-1451
This item has been discontinued and is no longer available. Please call customer service for assistance: 1-800-766-7000.
Alert:
TEFLON™ is a registered trademark of The Chemours Company FC LLC
Description
Description
  • Alnico magnet encapsulated in FDA Grade, USP Class VI Teflon™ PTFE low friction coating; durable and inert
  • Interrupted profile provides greater surface area and added turbulence when compared to the smooth surface of cylindrical bars
  • Pivot ring aids in reducing friction and chattering
  • Autoclavable
  • Manufactured in registered ISO 9001:2008 facilities, as verified by SGS certification
Specifications
Specifications
Yes
PTFE
0.31 in.
8mm
ISO 9001:2008
Red
Stir bar
Alnico Magnet