Fisherbrand™ 81-Place Polypropylene Cryo Storage Boxes

Polypropylene storage boxes provide compact storage and keep cryo vials organized on the bench, in the freezer, or during transport. Unlike cardboard models these sturdy boxes will not break down with exposure to moisture and heavy use.

 View more versions of this product

Catalog No. 14-081-F

  Check Availability
Add to Cart

    Specifications

    Color Assorted
    For Use With (Application) Provide compact storage and keep cryo vials organized on the bench, in the freezer or during transport
    For Use With (Equipment) 0.5, 1.5 and 2.0mL tubes and cryo vials
    Includes Clear translucent lids, matte panels on two sides, alphanumeric grid label
    No. of Places 81.0
    Temperature Range (English) -112° to 249.8°F
    Temperature Range (Metric) -80° to 121°C
    Material Polypropylene
    Product Type Cryo Storage Box
    Quantity 5 Pack

  • Description & Specifications
  • Promotions

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit