Learn More
Fisherbrand™ 96-Well Low-Profile, Skirted PCR Plates, Natural Available on GSA/VA Contract for Federal Government customers only.
Fit most thermal cyclers
 View more versions of this product
Catalog No. 14-230-238
Add to cart
Description
Description
Fit most thermal cyclers
  • Virgin polypropylene
  • Clear well bottom for sample visibility
  • Stackable
  • Raised-rim design
  • Sealable using adhesive films and foils, heat seals, and cap strips
  • 200µL maximum well volume when used with adhesive and heat seals
  • Cut-away corner and alphanumeric grid
  • SBS (Society for Biomolecular Screening) footprint
  • Automation friendly
  • Eight holes in the skirt aid plate positioning and removal from thermal cycler block
  • Cleanroom produced
This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.
Specifications
Specifications
Red
Polypropylene
Yes
200μL
PCR, RT-PCR applications
96.0
Skirted
Clear