Learn More
Fisherbrand™ Absorption Rectangular Quartz Flow Cells
Choose cells with quartz windows for transformation from the far ultraviolet through infrared
 View more versions of this product
Catalog No. 14-958-149
Description
Description
  • Compact size
  • All cells are 12.5mm square
  • Fused-on inlet and outlet tubes
  • With two clear windows and two black windows
  • Wavelength range for all cells is 190 to 2600nm and 2850 to 3600nm
Order Info
Select cells with center height appropriate to match the cell aperture center with the center position of the light beam in your spectrophotometer e.g. center height is 8.5mm for Beckman™ or Spectronic™ instruments, center height is 15mm for Hewlett-Packard™, Perkin-Elmer™, and Shimadzu™ instruments
Specifications
Specifications
Compact Flow Cell
Quartz
0.2 mm
35mm
12.4 μL
3.5mm
190 to 2600nm and 2850 to 3600nm
12.5mm