Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Fisherbrand™ Absorption Ultra Micro Rectangular Cells with PTFE Lid

Fit standard spectrophotometer cell holders

 View more versions of this product

Catalog No. 14-958-136



  Check Availability
Add to cart

Specifications

Description

  • All cells are 12.5mm wide
  • 1, 5 or 10mm path length rectangular cells
  • Reduce reagent use with Ultramicro cells
  • Quartz windows for transmission from far ultraviolet through the infrared
  • Glass windows for routine work in ultraviolet region and visible region

Description & Specifications

Specifications

100μL
For transmission from far ultraviolet through the infrared
45mm
Ea., 8 Ea./Cs.
Rectangular
12.5mm
2mm
Ultra Micro Absorption Cell
PTFE
Standard spectrophotometer cell holder
8.5mm
10mm
200 to 2500nm
2mm
Quartz
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit