Fisherbrand™ Activated Carbon Charcoal, 50 to 200 mesh
Activated Carbon 50 to 200 Mesh. For decolorizing oils, colored solutions, syrups. 300 to 75µm particle size. Not recommended for adsorbing solvents from gases
Quantity 4 lb.
 View more versions of this product
Catalog No. 05-690B
 N/A
N/A
This item has been discontinued and is no longer available. Please call customer service for assistance: 1-800-766-7000.  View substitute products »
    Chemical Identifiers
    Chemical Name or Material Fisherbrand™ Activated Carbon Charcoal, 50 to 200 mesh
    Specifications
    Quantity 4 lb.
    Chemical Name or Material Fisherbrand™ Activated Carbon Charcoal, 50 to 200 mesh
  • Description & Specifications
Related Products
Product Content Feedback