Learn More
Fisherbrand Adhesive Entryway Mats
Trap contaminating dirt, dust and bacteria before they invade cleanrooms
Description
Trap contaminating dirt, dust and bacteria before they invade cleanrooms
- Clear 2mil thick polyethylene sheets coated with environmentally friendly waterborne acrylic adhesive
- Pull-tab system makes it easy to peel off top sheet when it becomes soiled; remaining sheets stay intact and clean
- Will not curl, tear or wrinkle
- Adhesive will not transfer to footwear
- For standard tack needs: standard tack, 30; ideal for use in Electronic, Pharmaceutical, Laboratory and Food industry cleanrooms
Specifications
24 x 45 in. | |
2mil | |
Low-tack | |
40 clear sheets on white mat |