Fisherbrand™ Advanced Protection Disposable Facemasks
Provides convenient protection
 View more versions of this product
Catalog No. 17-100-898
  Check Availability
Add to Cart
    Specifications
    Thickness 3-ply
    Includes Ear loops
    Disposable Yes
    Color White
    Latex Free Yes
    Material Nonwoven Polypropylene (outer and inner) / Meltblown Polypropylene (filter)
    Size Universal
  • Description & Specifications
Related Products
Product Content Feedback