Learn More
Fisherbrand™ Aerosol-Barrier Tips Available on GSA/VA Contract for Federal Government customers only.
Prevent cross-contamination by using superior aerosol-barrier tips
 View more versions of this product
Catalog No. 02-707-150
  Check Availability
Add to cart
Description
Description
  • Unique small micron filter prevents aerosol contamination with no obstruction to airflow
  • Use aerosol barrier tips to ensure result integrity
  • Bulk packs are autoclavable; sterile products are not autoclavable
  • Certified by independent laboratory nonsterile; RNase, DNase, DNA, and pyrogen free
  • Natural color
This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.
Specifications
Specifications
Universal Fit Pipettor
5μL
Sterile
Yes
3.04cm
Universal Fit Pipettor
MicroPoint
Filtered
Natural
1.2 in.
DNase, RNase, DNA, and Pyrogen Free
Documents
Accessories