Fisherbrand™ Aerosol-Barrier Tips: 200–5000µL
Inert, hydrophobic filters prevent aerosol contamination
 View more versions of this product
Catalog No. 02-707-74
 N/A
N/A
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.  View substitute products »
    Specifications
    Packaging Lift-off Rack, 10 x 96
    Length (English) 2 in.
    Length (Metric) 5.08cm
    Sterility Sterile
    Autoclavable Yes
    Tip Style Aerosol Barrier, Large orifice
    Volume (Metric) 200μL
    Color White
  • Description & Specifications
Related Products
Product Content Feedback