Learn More
Fisherbrand™ Aerosol-Barrier Tips
Prevent cross-contamination by using superior aerosol-barrier tips
 View more versions of this product
Catalog No. 02-707-159
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
View substitute products »
Description
Description
  • Unique small micron filter prevents aerosol contamination with no obstruction to airflow
  • Use aerosol barrier tips to ensure result integrity
  • Bulk packs are autoclavable; sterile products are not autoclavable
  • Certified by independent laboratory nonsterile; RNase, DNase, DNA, and pyrogen free
  • Natural color
Packaging Code -- BP: Bulk Pack; RI: Rack Inserts; HR: Hinged Rack; HRS: Hinged Rack, Sterile; HR/T: Hinged Rack, Trace Metal-free; LR: Lift-off Rack; LRS: Lift-off Rack, Sterile; SR: Stack Rack; IWS: Individually Wrapped, Sterile.
Specifications
Specifications
Universal Fit Pipettor
100μL
Sterile
Yes
4.82cm
Universal Fit Pipettor
Bulk
Filtered
Natural
1.9 in.
DNase, RNase, DNA, and Pyrogen Free