Learn More
Fisherbrand Alkacid Full-Range pH Kit
pH accuracy to ±0.25
Catalog Number | Description | Price | Qty | Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Description | Price | Qty | Availability | |||||
A981
|
N/A | Range: 0.0 to 3.0; Blue |
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000. |
View substitute products » |
A985
|
N/A | Range: 10.5 to 12.5; Yellow |
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000. |
N/A |
A983
|
N/A | Range: 6.0 to 8.5; Yellow |
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000. |
View substitute products » |
A984
|
N/A | Range: 9.0 to 11.0; White |
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000. |
N/A |
Description
Provides sharp color change for each 0.5pH unit. Consists of A979 wide-range paper for determining approximate pH value and six specific short-range papers.
- Range 1 (pH 0.0 to 3.0)
- Range 2 (pH 3.5 to 5.5)
- Range 3 (pH 6.0 to 8.5)
- Range 4 (pH 9.0 to 11.0)
- Range 5 (pH 10.5 to 12.5)
- Range 6 (pH 12.0 to 14.0)
Order Info
Each paper comes in 15 ft.L x 0.25 in. W (4.6m x 6.4mm) roll with its own plastic dispenser. Appropriate color chart mounted inside. Seven rolls packed in hinged plastic box. Replacement short-range paper rolls available separately.
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.