Learn More
Fisherbrand™ Aluminum Weighing Dishes
Aluminum dishes for weighing and dispensing
 View more versions of this product
Catalog No. 08-732-101
Add to cart
Description
Description
Choose Fisherbrand™ Aluminum Weight Dishes with either smooth or fluted sides for weighing and dispensing.
With Fluted sides
  • Also can serve as evaporating dishes or dust covers Lightweight, yet sturdy Convenient finger-grip handle can be etched prior to weighing for identification
With Smooth sides
  • Lightweight dishes with flanged rims Slightly sloping sides facilitate stacking
  • Especially useful for materials that cannot easily be cleaned from dish surface
Specifications
Specifications
42mL
62 mm
Aluminum
With fluted sides
Weighing Dish
51 mm
17 mm
For weighing and dispensing