Learn More
Fisherbrand™ Amber Boston Round Bottles with White Polypropylene Pulp/Vinyl Lined Caps
Amber bottles are general use bottles ideal for when content integrity and UV protection is important. They ensure integrity for long term storage and offer excellent protection for UV-sensitive contents.
 View more versions of this product
Catalog No. 02-911-723
This item has been discontinued and is no longer available. Please call customer service for assistance: 1-800-766-7000.
View substitute products »
Specifications Description & Specifications
Specifications
4 oz., 125mL
4 oz.
Poly/Vinyl
Soda Lime Glass
22-400
125mL
PTFE
Amber
160/Cs.
Screw