Fisherbrand™ Amber Boston Round Bottles with White Polypropylene Pulp/Vinyl Lined Caps Available on GSA/VA Contract for Federal Government customers only.
Amber bottles are general use bottles ideal for when content integrity and UV protection is important. They ensure integrity for long term storage and offer excellent protection for UV-sensitive contents.
 View more versions of this product
Catalog No. 02-911-729
 N/A
N/A
This item has been discontinued and is no longer available. Please call customer service for assistance: 1-800-766-7000.  View substitute products »
    Specifications
    Capacity (English) 32 oz.
    Closure Screw
    Closure Material PTFE
    Closure Liner Poly/Vinyl
    Color Amber
    Material Soda Lime Glass
    Quantity 30/Cs.
    Capacity (Metric) 1000mL
    Closure Size 33-400
    Capacity 32 oz., 1000mL
  • Description & Specifications
Related Products
Product Content Feedback