Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Fisherbrand™ Amber Boston Round Bottles with Black Phenolic Rubber Lined Caps Available on GSA/VA Contract for Federal Government customers only.

For long term storage. Fisherbrand™ Amber Boston Round Bottles with Black Phenolic Rubber Lined Caps are ideal for use when content integrity and UV protection is important.

 View more versions of this product

Catalog No. 02-912-343


  Check Availability
Add to cart

Specifications Description & Specifications

Specifications

16 oz., 500mL
16 oz.
Rubber
Glass
Bottle, Boston Round
28-400
Narrow
Boston Round
480mL
Phenolic
Amber
12/Cs.
Black
Rubber
28-400
Screw
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit