Fisherbrand™ Amber Boston Round Bottles without Cap Available on GSA/VA Contract for Federal Government customers only.

Ideal for UV sensitive contents. Fisherbrand™ Amber Boston Round Bottles without Cap are for general use.

 View more versions of this product

Catalog No. 02-911-908

  Check Availability
Add to Cart

    Specifications

    Mouth Narrow
    Mouth Style Narrow
    Shape Boston Round
    Closure Included No
    Product Type Boston Round Bottle
    Neck Finish 20-400
    Quantity 288/Cs.
    Material Glass
    Closure Screw
    Capacity (Metric) 60mL
    Capacity (English) 2 oz.
    Color Amber
    Capacity 2 oz., 60mL

    • Features narrow screw neck opening, curved shoulders, and a round body
    • Glass offers minimal chemical interference with contents
    • Amber color absorb all wavelengths in the ultraviolet spectrum
    • Ensure integrity for long term storage and protection for UV-sensitive contents

  • Description & Specifications

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit