Learn More
Our pricing system is unavailable. You are viewing list price.
Please call 1-800-766-7000 to place your order or try our site again later.
Fisherbrand™ Amber Glass Jugs
Protect light-sensitive materials at an economical cost
 View more versions of this product
Catalog No. 02-912-276
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
Description
Description
  • Amber Type III soda-lime glass
  • Narrow mouth enables careful pouring and prevents spillage
  • 38mm-400 screw caps attached
Specifications
Specifications
130 oz., 3844.56mL
130 oz.
Polyethylene
Glass
Narrow
3844.56mL
Polyethylene
Amber
38-400
Screw