Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Fisherbrand™ Amber Wide Mouth Packer Bottles with Black PE Cone Cap Available on GSA/VA Contract for Federal Government customers only.

Ideal for storing dry materials. Fisherbrand™ Amber Wide Mouth Packer Bottles with Black PE Cone Cap are often used in chemical and environmental applications.

 View more versions of this product

Catalog No. 02-911-445


  Check Availability
Add to cart

Specifications

Description

  • Feature wide opening, making them ideal for general purpose use
  • Amber color filters out ultraviolet wavelengths, ideal for light sensitive solids
  • Classic "vitamin bottles" and are used in pharmaceutical packaging
Description & Specifications

Specifications

2 oz., 60mL
2 oz.
Polyseal Polyethylene Cone
Glass
Bottle, Wide Mouth Packer
33-400
Wide
Round
60mL
Phenolic
Amber
24/Cs.
Black
PolyCone
33-400
Screw
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit