Learn More
Fisherbrand™ Certified Cleaned Straight Sided Bottles
Ideal for soil sampling and environmental applications. Fisherbrand™ Certified Cleaned Straight Sided Bottles are manufactued from clear soda glass.
 View more versions of this product
Catalog No. 02-911-935
This item has been discontinued and is no longer available. Please call customer service for assistance: 1-800-766-7000.
View substitute products »
Specifications
Description
  • Cleaned per EPA guidelines for semi-volatiles, pesticides, PCBs and metals
  • Straight sided walls allow for the complete removal of contents
  • Bottles are autoclavable; caps are not
Description & Specifications
Specifications
1 oz., 30mL
1 oz.
Soda Lime Glass
432/PK.
28-400
Screw
30mL
Amber
432/cs.
No
Wide