Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Fisherbrand™ Amber Wide Mouth Packer Bottles without Cap

Ideal general purpose bottle for solids. Fisherbrand? Amber Wide Mouth Packer Bottles without Cap filters out ultraviolet wavelengths, making amber packers ideal for light sensitive solids.

 View more versions of this product

Catalog No. 02-911-936


  Check Availability
Add to cart

Specifications

Description

  • Wide mouth bottles allow for easy accessibility for removal of contents
  • Packer bottles are classic "vitamin bottles" and often used in pharmaceutical packaging, chemical and environmental applications
Description & Specifications

Specifications

2 oz., 60mL
2 oz.
Glass
216/Cs.
No
Wide
Round
60mL
Amber
216/cs.
Bottle, Wide Mouth Packer
33-400
33-400
Screw
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit