Fisherbrand™ Round Amber Wide Mouth Soda Lime Glass Packer Bottles without Cap
 View all options for this product
Catalog No. 02-911-798
This item has been discontinued and is no longer available. Please call customer service for assistance: 1-800-766-7000. 
    Capacity 0.5 oz., 15mL
    Capacity (Metric) 15mL
    Capacity (English) 0.5 oz.
    Closure Screw
    Closure Size 28-400
    Color Amber
    Material Soda Lime Glass
    Mouth Wide
    Packaging Type 624/Cs.
    Quantity 624/PK.
    Closure Included No
  • Description & Specifications
Related Products
Product Content Feedback