Fisherbrand™ ASTM™ Specific Gravity Hydrometers for Liquids Heavier than Water, 275mL Liquid Volume Available on GSA/VA Contract for Federal Government customers only.

Accuracy meets or exceeds ASTM E 100 specifications

 View more versions of this product

Catalog No. 11-6037L

  Check Availability
Add to Cart

    Specifications

    Specific Gravity Range 1.4 to 1.45
    Certifications/Compliance ASTM E126
    For Use With (Application) Liquids heavier than Water
    Volume (Metric) 275mL
    Temperature of Standardization 60°F
    ASTM Number 119H
    Increments 0.0005
    Length (Metric) 330mm

  • Description & Specifications

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit