Learn More
Fisherbrand™ ASTM™ Specific Gravity Hydrometers for Liquids Lighter than Water
Accuracy of hydrometer meets or exceeds ASTM™ E 100 specifications.
 View more versions of this product
Catalog No. 11-603-3XC
Add to cart
Description
Description
Accuracy of hydrometer meets or exceeds ASTM™ E 100 specifications.
  • Use Cylinder 08-530K
  • Temperature of standardization: 60°F
Specifications
Specifications
107H
3-point Certification, ASTM E126
Liquids heavier than Water
260mL
0.001
0.001
260mm
0.9 to 0.95
60°F