Learn More
Fisherbrand™ ASTM™ Specific Gravity Hydrometers for Liquids Lighter than Water
Accuracy of hydrometer meets or exceeds ASTM™ E 100 specifications.
Description
Accuracy of hydrometer meets or exceeds ASTM™ E 100 specifications.
- ASTM API 260mm ASTM Number: 104H 0.001 0.750 to 0.800 60°F Plain Form Hydrometer
- W/3-pt certification
Order Info
A certificate of calibration is available; simply add the letter “C” to the end of the catalog number when ordering.
Compliance
Calibrated according to ASTM™ E 126 specifications
Specifications
Hydrometer | |
3-point Certification, Cited in ASTM E100-14 Table 1 | |
0.750 to 0.800 ASTM 104H-62 | |
60°F |
104H | |
Petroleum products | |
260 mL |