Learn More
Fisherbrand™ ASTM™ Specific Gravity Hydrometers for Liquids Lighter than Water
Accuracy of hydrometer meets or exceeds ASTM™ E 100 specifications.
 View more versions of this product
Catalog No. 11-603-4AC
Add to cart
Description
Description
Accuracy of hydrometer meets or exceeds ASTM™ E 100 specifications.
  • Use Cylinder 08-530K
  • Temperature of standardization: 60°F
Specifications
Specifications
82H
3-point Certification, ASTM E126
Liquids heavier than Water
275mL
0.0005
0.0005
330mm
0.65 to 0.7
60°F