Learn More
Fisherbrand™ ASTM™ Specific Gravity Hydrometers for Liquids Lighter than Water
SDP
Accuracy of hydrometer meets or exceeds ASTM™ E 100 specifications.
 View more versions of this product
Catalog No. 11-603-4FC
Add to Cart
Description
Description
Accuracy of hydrometer meets or exceeds ASTM™ E 100 specifications.
  • Use Cylinder 08-530K
  • Temperature of standardization: 60°F
Order Info
A certificate of calibration is available; simply add the letter “C” to the end of the catalog number when ordering.
Compliance
Calibrated according to ASTM™ E 126 specifications
Specifications
Specifications
87H
3-point Certification, ASTM E126, Cited in ASTM E100-14 Table 1
Soda Lime Glass
0.900 to 0.950 ASTM 87H-62
60°F
0.0005
330 mm
Liquids heavier than Water
275 mL
Product Suggestions
Product Suggestions