Fisherbrand™ ASTM™ Thermometers for Flash Point Testing

Guaranteed to meet ASTM E 1 specifications in every detail

 View more versions of this product

Catalog No. 13-501

 N/A
N/A

This item has been discontinued and is no longer available. Please call customer service for assistance: 1-800-766-7000. 

Alert

The products indicated contain mercury. Do not place products containing mercury in the trash; they are to be recycled or disposed of as hazardous waste. Products containing mercury may be regulated by law and may not be available in all countries.


    Specifications

    For Use With (Application) Pensky-martens, tag open, tag closed
    Immersion Length 57mm
    Length (English) Overall 11.37 in.
    Length (Metric) Overall 290mm
    Subdivision 0.5°C,0.5°C
    Temperature Range (Metric) -5° to +110°C
    Certifications/Compliance ASTM E 1

  • Description & Specifications

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit